Learn More
Medchemexpress LLC HY-15290 5mg Medchemexpress, AIM-100 CAS:873305-35-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY152905MG
Encompass_Preferred
Medchemexpress, HY-15290 5mg AIM-100 CAS:873305-35-2 AIM-100 is a potent and selective Ack1 inhibitor with an IC50 of 21.58 nM. AIM-100 also inhibits Tyr267 phosphorylation. AIM-100 does not inhibits other kinases including PI3-kinase and AKT subfamily members. AIM-100 has an anticancer effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1698