Learn More
Across International Ai GL14 Chilling Fluid or Vacuum Connectors - Pack of 5
SDP
Supplier:  Across International GL14X5
Encompass_Preferred
GL14 chilling fluid or vacuum connector with sealing gasket, pack of 5. Fits some models of our glass condensers of SolventVap rotary evaporators and glass jacketed reactors.
Catalog No. NC1955141