Learn More
Across International Ai 2L SolventVap w/ Electric Lift and Cold Trap Condenser 110V
SDP
Supplier:  Across International SE05C.110
Encompass_Preferred
Ai SolventVap 2L (evaporating flask capacity) series rotary evaporators come with a modular heating bath, vertical glassware, and digital temperature/rotation control. Its optimized bath volume allows for rapid heat-up times. Thats why our SolventVap evaporators consistently deliver reliable and reproducible process results that are usually reserved for more costly systems. Another important aspect of the SolventVap is the safety features, which includes dry-run protection, water bath over-temperature shut-off protection, and fuse protection.
Catalog No. 50-225-1289