Learn More
Ahlstrom Filter Papers™ – Grade 617 PROMO
Supplier:  Ahlstrom 61700900PM
 View more versions of this product
Catalog No. 0979066APM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
35 μm
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing
9 cm
Fast
617
Product Suggestions
Product Suggestions