Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 617
An extremely fast filtering grade that has a larger particle retention than Grade 631. Grade 617 is a thicker filter paper with higher loading capacity used for rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions.
Supplier:  Ahlstrom-Munksjo 61703300
 View more versions of this product
Catalog No. 09-790-66H
Add to Cart
Description
Description
Specifications
Specifications
35 μm
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing
33 cm
Fast
617
Product Suggestions
Product Suggestions