Learn More
Ahlstrom-Munksjo Standard Non Wet-Strengthened Qualitative Filter Paper, Grade 617
An extremely fast filtering grade that has a larger particle retention than Grade 631. Grade 617 is a thicker filter paper with higher loading capacity used for rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions.
Supplier: Ahlstrom-Munksjo 61701850
Specifications
35 μm | |
Rapid filtration of coarse particles and gelatinous precipitates such as in oils and resin solutions; construction product testing | |
18 cm |
Fast | |
617 |