Learn More
Ahlstrom™ Filter Papers – Grade 615 PROMO
Supplier: Ahlstrom 61500750PM
Specifications
25 μm | |
Filtering aluminum, ferric, magnesium hydroxides. For construction filtration work, filtration of insoluble iron and ore. | |
7.5 cm |
Fast | |
615 |