Learn More
Ahlstrom™ Filter Papers – Grade 601 PROMO
Supplier:  Ahlstrom 60101100PM
Catalog No. 0979055EPM
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
2.5 μm
Clarifying liquids, rapid filtration of coarse precipitates: lead sulfate, lead chromate, calcium oxalate/carbonate, aluminum phosphate
11 cm
Medium
601
Product Suggestions
Product Suggestions