Learn More
Scientific Instrument Services Inc Agilent/HP 5973 Source Heater
Supplier: Scientific Instrument Services Inc HPC5
HPC5 is a direct replacement for the heater cartridge on the 5973/75 source block.