Learn More
Axygen™ Agarose LM
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
Axygen™ Low melting agarose allows for the recovery of undamaged nucleic acids below their denaturation temperature.
$411.00 - $4120.00
Specifications
Product Type | LM Agarose Gel |
---|---|
Dnase Free | Yes |
For Use With (Application) | Tissue and cell culture and viral plaque assays |
Material | Agarose |
Melting Temperature | ≥76°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-223-083
![]() |
Axygen™
AGRLM100 |
100 g |
Case for $411.00
|
|
|||||
14-223-042
![]() |
Axygen™
AGRLM501 |
10 x 50 g |
Case of 10 for $2,450.00
|
|
|||||
14-223-043
![]() |
Axygen™
AGRLM1001 |
10 x 100 g |
Case of 10 for $4,120.00
|
|
|||||
Description
The low gelling temperature ensures that the agarose will be in a liquid state at a temperature range where In-Gel manipulations can be performed without prior extraction of the DNA from the gel slice. Low Melting (LM) Agarose is derivatized by organic synthesis which generates methoxylate groups from the basic agarose structure. Agarose LM is also optimized for preparative electrophoresis and isolation of DNA fragments.
- Highest gelling/melting temperatures and gel strength
- Electrophoresis of DNA fragments ≥1000bp
- Suitable for tissue and cell culture and viral plaque assays
- Ideal for preparative electrophoresis
Specifications
LM Agarose Gel | |
Tissue and cell culture and viral plaque assays | |
≥76°C | |
15° to 25°C |
Yes | |
Agarose | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.