Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agarose Gel-Loading Dye, 6X (Contains 15% Ficoll™ 400/Molecular Biology), Fisher BioReagents
Description
Poly Bottle; 5mL
This tracking dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.Specifications
Agarose Gel-Loading Dye, 6X | |
Liquid | |
915-67-3, 26873-85-8 | |
DNase free | |
Amaranth (<1.0%), Ficoll™ (15%), Proprietary Component (<1.0%), and Water. |
0.3 to 0.6 AU at 252nm, 0.5 to 0.8 AU | |
Contains 15% Ficoll™ 400 | |
5 mL | |
Molecular Biology | |
Poly Bottle |
Safety and Handling
Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction