Learn More
Vector Laboratories Sambucus Nigra Lectin (SNL, SNA, EBL) Agarose Bound 2ml
Supplier: Vector Laboratories AL13032
Sambucus nigra lectin (SNL) is isolated from elderberry bark and binds preferentially to sialic acid attached to terminal galactose in a-2,6 and to a lesser degree, a-2,3 linkage. Also called SNA (Sambucus nigra agglutinin) and EBL (elderberry lectin), SNL can inhibit cell-free protein synthesis. This agarose-bound SNL is prepared using affinity-purified lectin and heat stable, cross-linked 4% agarose beads. The attachment of the lectin through a hydrophilic spacer arm to the beads is carefully controlled to preserve the activity of and minimize conformational changes to the bound lectins. The recommended inhibiting/eluting sugar is 500 mM lactose in buffered saline followed by 500 mM lactose in acetic acid.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.