Learn More
Nelson Jameson Company AGAR STRIPS RCS AIR SMPLR 50PK
Supplier: Nelson Jameson Company 0803032 BF
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Agar strips for the RCS Air Sampler. 50 strips per box. Rose Bengal. Use for yeasts and mold.