Learn More
Thermo Scientific AG14 GUARD COLUMN 4MM
Supplier: Thermo Scientific 046134
IonPac AG14 Guard Column, Dimensions:4 x 50 mm, Maximum Operating Pressure: 27.6 MPa (4000 psi), Mobile Phase Compatibility: pH 2-11; 0-100% HPLC solvents,Bead Diameter ( micrometer): 9.0 micrometer, Pore Size: 100 angstrom, Crosslinking (%