Learn More
Enzo Life Sciences Antigen Retrieval Reagent, pH 6 (10X) (100ml)
Supplier: Enzo Life Sciences ENZACC1120100
Antigen Retrieval Reagent, pH 6 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin. Formulation: Liquid. In citrate buffer. Long Term Storage: +4°C. Applications: IHC .