Learn More
World Precision Instrument Ag/AgCl Electrode-EP08
Supplier: World Precision Instrument EP08
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.