Learn More
Bioanalytical Systems Inc AG/AGCL (3M KCL) REFERENCE ELE
Supplier: Bioanalytical Systems Inc MF2056
RE-5B silver/silver chloride (3M potassium chloride) reference electrode with flexible connector. 7.5 cm long glass body and membrane with 6 mm outer diameter.