Learn More
Enzo Life Sciences AG-879 (25mg). CAS: 148741-30-4
Supplier: Enzo Life Sciences BMLEI2580025
AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells. Purity: ≥99% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.