Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ Afinion™ HbA1c Test Dx
A test for quantitative determination of Glycated Hemoglobin (HbA1c) in Human Whole Blood; Used to monitor metabolic control in patients with diabetes. Afinion™ HbA1c Dx Test can also aid in the diagnosis of diabetes or to help identify patients who may be at risk for developing the disease.
Supplier: Abbott™ 1116794
This item is not returnable.
View return policy
Description
- Immediate point of care HbA1c results for efficient monitoring of glycemic control
- Ready-to-use controls in two levels
- Capillary and venous whole blood sample material
- 1.5 μL sample volume
- Measuring range: 4% to 15% HbA1c
- Storage: Up to 3 months at Room Temperature
Specifications
Up to 3 months (at Room Temperature) | |
Capillary Blood or Anticoagulated Venous Blood (EDTA) | |
Traceable to the IFCC Reference Method | |
4% to 15% HbA1c | |
3 min. |
NGSP and IFCC Certified | |
HbA1c Dx In Vitro Diagnostic Test | |
15/Pk. | |
1.5 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title