Learn More
Jackson Immuno Research Labs AffiniPure Alpaca Anti-Rabbit IgG (H+L) | 1.0mg
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Jackson Immuno Research Labs 611005215
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Target: Rabbit | Host: Alpaca | Antibody Format: Whole IgG | Specificity: IgG (H+L) | Minimal Cross Reactivity: Bovine, Human, Mouse Serum Proteins | Conjugate: Unconjugated