Learn More
MSA™ Advantage™ 4000 Twin-Port Facepiece
Two bayonet cartridge adapters mount directly onto the facepiece lens for compatibility with all Advantage cartridges and filters
Supplier: MSA™ 10108568
This item is not returnable.
View return policy
Description
- Approved with all MSA Advantage Cartridges
- Features two mounting locations for cartridges or filters on the right and left sides
- Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
ANSI Z87.1 2003, NIOSH 42 CFR Part 84 | |
Twin-Port Facepiece | |
Msa™ respirator models | |
4-point |
Rubber | |
Medium | |
Dupont™ Kevlar™ aramid fiber net |