Learn More
MSA™ Advantage™ 4000 Twin-Port Facepiece
Two bayonet cartridge adapters mount directly onto the facepiece lens for compatibility with all Advantage cartridges and filters
Supplier:  MSA™ 10108567
 View more versions of this product
Catalog No. 17-200-087
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Approved with all MSA Advantage Cartridges
  • Features two mounting locations for cartridges or filters on the right and left sides
  • Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
Specifications
ANSI Z87.1 2003, NIOSH 42 CFR Part 84
Twin-Port Facepiece
Respiratory applications
Kevlar net, four-point
Silicone, Rubber
Medium
Msa™ respirator models
Nosecup, bayonet adapters in lens, facepiece elastomers
Product Suggestions
Product Suggestions