Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Advanced TC treated 6-well Cell Culture Microplates
For high-throughput screening in cell based test systems.
Supplier: Greiner Bio-One 657960
Description
- Non-pyrogenic, non-cytotoxic
- Improved cell adherence
- Consistent cell attachment
- Homogenous cell growth
- In-vivo like morphology
- Increased cell yield
- Permits usage of serum-reduced or serum free media
Specifications
Clear | |
Polystyrene | |
Flat | |
Clear | |
Polystyrene | |
Sterile | |
Cell Culture Microplate | |
6 | |
Lid | |
1 | |
128 x 85 x 19 mm | |
85.48 mm |
Closure Included | |
Not Autoclavable | |
5 mL (Well) | |
9.6 sq. cm | |
Rectangular | |
Tissue Culture | |
2 to 5 mL | |
Yes | |
100 | |
Yes | |
127.76 mm | |
19 mm |
Safety and Handling
ShelfLife : 2 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction