Learn More
Perkin Elmer US LLC Advanced Platform THGA Graphite Tubes with End-Cap, Pkg. 5
Supplier: Perkin Elmer US LLC N9307832
The exclusive and patented Advanced Platform design offers an alternative to our Standard Integrated Platform design. With the current Integrated Platform design, the platform attaches to the tube wall along a single side that runs the length of the platform.