Learn More
Perkin Elmer US LLC Advanced Platform HGA Graphite Tubes, Pkg. 20
Supplier: Perkin Elmer US LLC N9307835
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The exclusive and patented Advanced Platform design offers an alternative to our Standard Integrated Platform design. With the current Integrated Platform design, the platform attaches to the tube wall along a single side that runs the length of the platform.