Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Advanced Instruments 621 Cryoscope Standard Bottle
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Standard bottle used when calibrating and checking the performance of cryoscopes
Supplier: Advanced Instruments 3LA032
This item is not returnable.
View return policy
Description
- Capacity: 110mL (3.7 oz)
- Expected Value: 621 m°H (-600 m°C)
- Expected Range: 619 - 623 m°H (-598 to -602 m°C)
Specifications
Calibration Standard Bottle | |
Cryoscope, osmometers and other calibration equipment | |
110mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction