Learn More
Epoch Life Science Inc GenCatch™ Advanced Gel Extraction Kit (50 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
Supplier: Epoch Life Science Inc 2260050
Agarose gel DNA extraction kit. Silica membrane spin column based. Sample volume: 850 ul. Column binding capacity: 40 ug DNA. Minimum elution volume: 20ul. 18 month shelf life.