Learn More
Cole-Parmer Advantec Narrow Range pH Paper Booklet, Bromophenol blue (2.8-4.4 pH), 200/bx
Supplier: Cole-Parmer 5920105
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These pH test paper booklets feature narrow pH unit graduation (0.2 to 0.4 pH) and can be used for severe pH measuring. Each leaflet is 1/4" x 2-3/4" (7 x 70-mm). There are 20 leaflets per booklet and 10 booklets per box.