Learn More
Sciteck™ AdultaCheck™ Adulteration Test Strips
The first dry chemistry test strip to ever test for creatinine and glutaraldehyde in urine
$66.00
Specifications
For Use With (Application) | For reagent impregnated test pads for specific analytes |
---|
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Price | Quantity & Availability | ||||
23-424-4508
|
Sciteck™
30150 |
Creatinine, Nitrite, Chromate, Oxidants, Glutaraldehyde, Ph | 50 Tests |
Pack of 50 for $66.00
|
|
||||
Description
Sciteck Specimen Validity Testing (SVT™)
- In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
- Adulteration testing products to include extensive proprietary SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line On-site adulteration testing
Specifications
For reagent impregnated test pads for specific analytes |