Learn More
Fisherbrand™ Adjustable Volume Dispenser Bottle LDPE/PMP
SDPSCI_ED
Maximum volume, 1000 mL
Catalog No. S37104
Add to Cart
Description
Description
  • Graduated clear PMP measuring cup
  • Polyethylene tube and bottle for easy adjustment
  • Variation in height determines the volume to be dispensed
Specifications
Specifications
Dispenser Bottle
Polyethylene Tube, Polyethylene Bottle
1000 mL (Max.)
Education
Product Suggestions
Product Suggestions