Learn More
Eisco™ Adjustable Clear Safety Goggles, Vented
Click to view available options
Adjustable Clear Safety Goggles, Vented
Description
- Fit over most prescription lens
- Includes side vents to prevent lenses from fogging up
- Features black adjustable strap to fit most
Warranty and Services
1 year limited manufacturer warranty
Specifications
Lens Type | Splash protection |
Lens Tint | Clear |
For Use With (Application) | Safety ware |
No. per Pack | 1 Ea. |
Size | Universal |
Material | Polycarbonate |