Learn More
Invivogen Adju-Phos adjuvant, 250 mL
Supplier: Invivogen VACPHOS250
Adju-Phos adjuvant is an aluminium phosphate wet gel suspension. It induces a Th2 response by improving the attraction and uptake of antigen by antigen-presenting cells (APCs). It can also activate innate immunity pathways triggered by pattern recognition receptors (PRRs).