Learn More
Advanced ImmunoChemical, Inc. Adiponectin, Source - Pooled Human Plasma, Calibrator, Standard In Immunoassays, 0.05mg
Supplier: Advanced ImmunoChemical, Inc. 8ADP0.05 MG.
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc.
Antigens / Metabolic Syndrome / Adiponectin
Source: Pooled human plasma. Blood sample from the tissue donor was tested and found negative for HBsAg, HCV, HIV-1 and 2 antibodies, HCV antibodies, and syphilis; Applications: Calibrator or standard in immunoassays
Analysis: Purity >95% (SDS-PAGE); Immunological activity: Immunoreactivity is checked using monoclonal antibodies specific to human adiponectin; Purification: Affinity and hydrophobic interaction chromatography; Presentation: Lyophilized from solution in 25 mM Tris-HCl, pH 7.5, 0.1% CHAPS. Product is strongly recommended to reconstitute with 25 mM Tris-HCI, pH 7.5, 0.1% CHAPS to prevent any loss of immunoreactivity; Storage Conditions: Before and after reconstitution at -20 °C (between -15 and -30 °C allowed); Other information: Avoid repeated freezing and thawing. Aliquot the product after reconstitution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.