Learn More
Medchemexpress LLC HY-W017522 500mg Medchemexpress, Adipic acid CAS:124-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW017522500MG
Encompass_Preferred
Medchemexpress, HY-W017522 500mg Adipic acid CAS:124-04-9 Adipic acid is found to be associated with HMG-CoA lyase deficiency, carnitine-acylcarnitine translocase deficiency, malonyl-Coa decarboxylase deficiency, and medium Chain acyl-CoA dehydrogenase deficiency, which are inborn errors of metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3413