Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel QuickVue™ AdenoPlus™ External Controls
AdenoPlus external controls are to be used with the AdenoPlus test only andare intended to verify that the test reagents are working and that the test isperformed correctly.
Supplier: Quidel RPSADSTD
Description
Positive Control
- Buffered solution containing detergent and recombinant adenovirus hexon protein as well as additional proteins to
simulate biological matrix
Negative Control
- Buffered solution containing detergent and proteins to simulate biological matrix
Specifications
Room temperature not to exceed 86°F(30°C) | |
Adenoviral Conjunctivitis External Controls | |
1 vial labeled positive control and 1 vial labeled negative control |
Positive and Negative | |
A rapid, immunoassay test for the visual, qualitative, in vitro detection of Adenoviral antigens (hexon protein)directly from human eye fluid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction