Learn More
Perkin Elmer US LLC 1⁄4 in. (6.4 mm) Internal Screw Thread Adapter with 0.7 mm O.D. Pt/lr Capillary Nipple, Type F
Supplier: Perkin Elmer US LLC B0193873
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. Type F in instrument manual diagrams.