Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Adapter for Eppendorf™ Conical Tubes with Screw Cap
Adapter for 25 mL conical tubes with screw cap for rotors for conical 50 mL tubes
Supplier: Eppendorf™ 5820734000
This item is not returnable.
View return policy
Description
Specifications
Adapter | |
50 mL Tubes | |
50 mL |
Conical | |
For rotors for conical 50 mL tubes | |
Set of 6 pieces |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction