Learn More
Chemglass Life Sciences Adapter Cord, 14", from Module to Standard Plug
SDP
Supplier:  Chemglass Life Sciences CLS143819A
Encompass_Preferred
Adapter Cord, 14", from Module to Standard Plug. The Chemglass Life Science power cord is 6: IEC 60320-1 to NEMA 5-15. The adapter cord is 14 and adapts from a male IEC to a female u-ground plug. Adapter Cord only. Pictured is the CLS-1438-002 pH controller.
Catalog No. 50-153-2213