Learn More
Chemglass Life Sciences Adapter, Connecting Vial, 14/20, Glass Only
Supplier: Chemglass Life Sciences CG1206W245
Adapter, Connecting Vial, 14/20, Glass Only. Accessory for the CG-1206-W allihn condenser.