Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activator Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.5M ETT in Acetonitrile
$13904.00 - $48022.00
Specifications
CAS | 89797-68-2, 75-05-8 |
---|---|
Boiling Point | 82°C.The physical data is that of the main component. |
Flash Point | 13°C |
Packaging | Returnable Steel Drum |
Physical Form | Liquid, clear |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-090-032
|
Honeywell Chemicals
BC72556 |
56 L |
Each for $13,904.00
|
|
|||||
60-090-030
|
Honeywell Chemicals
BC725200 |
200 L |
Each for $48,022.00
|
|
|||||
Description
Specifications
89797-68-2, 75-05-8 | |
13°C | |
Liquid, clear |
82°C.The physical data is that of the main component. | |
Returnable Steel Drum | |
Activator reagents |
Safety and Handling
Recommended Storage : Room Temperature
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title