Learn More
Biotium Activated Charcoal Decontamination Bag
Supplier: Biotium 22007
Activated charcoal retains ethidium bromide, GelRed and GelGreen efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.