Learn More
Sigma Aldrich Fine Chemicals Biosciences Activated charcoal DARCO(R), 12-20 mesh, granular | 7440-44-0 | MFCD00133992 | 250G
Supplier: Sigma Aldrich Fine Chemicals Biosciences 242241250G
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Activated charcoal DARCO(R), 12-20 mesh, granular | Mol Wt: 12.01 | 7440-44-0 | MFCD00133992 | 250G