Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell ActivArmr™ 23-491 PVC-Coated Hi-Vis Gloves with Foam Insulation
Click to view available options
10
Description
- Tough PVC vinyl coating offers abrasion resistance
- Coating remains soft and flexible in low temperatures
- Cotton jersey liner increases user comfort
- Foam insulation keeps hands warm in cold applications
- Gloves are repellent to water, oil, and grease
- Enhanced grip when handling heavy-duty objects
- Hi-vis color is clearly visible in low light conditions
- Curved finger design with a wing thumb
Specifications
ANSI Abrasion Rating | ANSI 4 |
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009; Category II, REACH Compliant |
Construction | Cut & Sewn |
Coating Material | PVC |
Color | Hi-vis Orange |
Cut Resistance Rating | ANSI A2 |
Hand | Specific |
Latex Free | Yes |
Size | 10 |
EN388 Standard | 3121B |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction