Learn More
ECM Bioscience Actin Filament Regulation Immunocytochemistry Kit, ECM Biosciences
Supplier:  ECM Bioscience AK7600
Encompass_Preferred
The actin filament regualtion kit can be used to compare labeling between actin filaments and total actin. The kit contains two antibodies for detecting total actin, and Phalloidin:FITC for labeling actin filaments. In addition, secondary reagents conjugated to DyLight 594 are included for dual labeling experiments with Phalloidin:FITC. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms, Ck
Catalog No. 50-206-8604