Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Acrylic Storage Modules for Sample Cups



Storage for hundreds of 13mm sample cups in less than a square foot of space.
Specifications
Type | Storage Module |
---|---|
Material | Acrylic |
Color | Clear |
Description | Storage rack for sample cups |
Catalog Number | For Use With | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | For Use With | Price | Quantity & Availability | ||||||
S29192
![]() ![]() |
100 Sample Cups | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
S29193
![]() ![]() |
Storage Holder: 14 810 67 | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Clear acrylic shelves with white base
- Solid cover available separately to protect top holder
- Up to six holders can be stacked under one solid cover
Order Info
Racks with colored bases are available on special order at slightly higher prices. Contact your Fisher Scientific Customer Service Representative for ordering information.
Specifications
Storage Module | |
Acrylic | |
Clear | |
Storage rack for sample cups |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title