Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Acrylic Sign: WARNING: HAZARDOUS VOLTAGE INSIDE. CAN SHOCK, BURN, OR CAUSE DEATH. … (English/Spanish)
Improve workplace safety with proper signage and markings. This bilingual Brady™ acrylic sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$14.50
Specifications
Material | Acrylic |
---|---|
Shape | Rectangular |
Dimensions (L x W) | 10.75 x 4.5 in. |
Displayed Text | WARNING: HAZARDOUS VOLTAGE INSIDE. CAN SHOCK, BURN, OR CAUSE DEATH. … (English/Spanish) |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-109-685
|
Brady™
46348 |
Black |
Each for $14.50
|
|
|||||
19-109-686
|
Brady™
46349 |
Red on White |
Each for $14.50
|
|
|||||
Description
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Acrylic | |
Rectangular | |
10.75 x 4.5 in. | |
WARNING: HAZARDOUS VOLTAGE INSIDE. CAN SHOCK, BURN, OR CAUSE DEATH. … (English/Spanish) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title