Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Acrylic High Vacuum Pump Plate
This affordable quality high vacuum pump plate can accommodate bell jars up to 9 inches in diameter.
Supplier: Fischer Technical Company™ APRUBGAS
Description
- Made of acrylic and does not require reinforcing ribbons, which limit the size of bell jars you can use
- The 0.38 in. hose barb connection is compatible with most vacuum pumps
- The 10.75 x 1.5 in. base has four suction cups to hold the plate in place
- Replacement gaskets available
Specifications
9 in. dia. Bell Jar | |
Replacement Seal |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction