Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stoelting™ Acrylic Brain Matrices
Stoelting™ Acrylic Brain Matrices are precisely machined and highly polished to ensure reproducible sectioning.
Supplier: Stoelting™ 51381
Description
- Rat and mouse brain matrices are available
- Coronal and sagittal sections at 1 mm intervals
- All coronal brain matrices have an additional mid-line sagittal cut to aid in the separation of the left and right hemispheres.
- Mouse brain matrices can also be used on neonatal rat brains. Divisions are 1 mm apart.
Specifications
Neuroscience | |
40 to 75 g | |
Mouse, Rat | |
Sagittal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction