Learn More
Andwin Scientific Acro 50 Vent Devices with PTFE Membrane - 0.2 um, 1/8 in. MNPT, 18/pk
SDP
Supplier:  Andwin Scientific 4400
Encompass_Preferred
Venting bioreactors, fermentation tanks, and carboys. Sterile gas purging of culture vessels. Filtering aggressive solvents.
Catalog No. NC0890651