Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acridine Orange Hemizinc Salt, Fisher BioReagents™
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass
Description
Acridine Orange is suitable for microscopy and as a specific stain for DNA and RNA.
Specifications
Melting Point | 165°C |
Extinction Coefficient | 1100g(-1) cm(-1) min. |
Identification | Pass Test |
Lambda Max | 488 to 492nm |
Formula Weight | 369.94 |
Loss on Drying | 5% max. |
Packaging | Amber Glass |
Color Index No. | 46005 |
Color | Orange |
Quantity | 10 g |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction